Ampicillin sodium salt for tissue culture, sterile, Calbiochem®

Synonyms: D(-)-α-Aminobenzylpenicillin Sodium Salt

171255-20ML
CA80055-788
Ampicillin sodium salt for tissue culture, sterile, Calbiochem®
Ampicillin sodium salt

Sterilized by gamma-irradiation


Kills growing cells by interfering with the terminal reaction in bacterial wall synthesis. Reconstitute in 20mL sterile deionized water to prepare a 10mg/mL solution. Potency (anhydrous): >=845µg/mg.

Formula: C₁₆H₁₈N₃NaO₄S
MW: 371.39
MDL Number: MFCD00064313
CAS Number: 69-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR