Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Contour™ Laboratory Tables

Supplier: VWR International
VWR® Contour™
CFTM3630R30D CFT-4830-A CFT-4836-A VFT-3630-DRT VFT-7230-DRT CFTM4830R36D VFT-4830-DRT CFT-3630-A CFTM4830R30D CFT-7236-A CFT-3636-A CFT-7230-A CFTM7230R30D CFTM3630R36D CFTM7230R36D
Made with welded leg assemblies connected to steel apron rails with metal fasteners, Contour laboratory tables provide a stable work surface.

  • Light neutral color

The rails for the Contour laboratory tables are 10.2 cm (4") high and formed of 16-gauge steel

The resin worktop, molded from black epoxy resin with no laminates, will not bubble or crack. Because the resin is inert, this worktop is highly resistant to acids, reagents, and other chemical reactions. The durable, economical resin worktop is smooth, self-extinguishing, heat-resistant, contains no asbestos, and has a low-glare surface that is easy to clean and maintain.

Table legs are made of 5.1 cm (2") square welded tubing to provide a stable, solid workplace. Leg assemblies include leveling bolts, stretchers, and stringers for better support. Lockable casters are available.

Ordering information: Table frames and table tops are sold separately.



 

ORDER

Expand All / Collapse All

Frames
Description Depth Height Width No. of Drawers Supplier No. VWR Catalog Number Unit Price Quantity
Sitting Height Table Frame 30" 30" 36" 1   CFT-3630-A Each Retrieving
Product available on GSA Advantage®
Sitting Height Table Frame 30" 30" 48" 1   CFT-4830-A Each Retrieving
Product available on GSA Advantage®
Sitting Height Table Frame 30" 30" 72" 2   CFT-7230-A Each Retrieving
Product available on GSA Advantage®
Sitting Height Table Frame 30" 30" 36" 1   CFTM3630R30D Each Retrieving
Product available on GSA Advantage®
Standing Height Table Frame 30" 36" 36" 1   CFT-3636-A Each Retrieving
Product available on GSA Advantage®
Standing Height Table Frame 30" 36" 48" 1   CFT-4836-A Each Retrieving
Product available on GSA Advantage®
Standing Height Table Frame 30" 36" 72" 2   CFT-7236-A Each Retrieving
Product available on GSA Advantage®
Standing Height Table Frame 30" 36" 48" 1   CFTM4830R36D Each Retrieving
Product available on GSA Advantage®

Worktops
Description Depth Thickness Width Supplier No. VWR Catalog Number Unit Price Quantity
Resin Worktop 30" 1" 36" VFT-3630-DRT VFT-3630-DRT Each Retrieving
Resin Worktop 30" 1" 48" VFT-4830-DRT VFT-4830-DRT Each Retrieving
Resin Worktop 30" 1" 72" VFT-7230-DRT VFT-7230-DRT Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Width Supplier No. VWR Catalog Number Unit Price Quantity
Casters, Set of 4 353/4"   CFSP-0045-04 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty