AK225 Solvents, Techspray™
Supplier: ITW Chemtronics
1665-8S
14216-050CS 522.55 USD
14216-050
AK225 Solvents, Techspray™
These AK225-based solvents provide excellent solvency and can be used for vapor degreasing, benchtop cleaning, and aerosol applications
Kleen ATMS azeotropic-blend solvent is an ideal 1,1,1-trichloroethane replacement. Flux Remover AMS solvent is designed for defluxing via vapor degreasing or cold-batch cleaning. Rapid evaporation. Nonflammable.
Ordering information: For an optional brush attachment designed for use with the aerosol spray cans, see 14216-118.