MR-VP Medium, Dehydrated, HiMedia Laboratories
Supplier: HiMedia Laboratories
GM070I-500G M070I-500G
95020-178EA 101.54 USD
95020-178 CA95020-178
MR-VP Medium, Dehydrated, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For studying methyl red and voges proskauer tests to differentiate amongst coli-aerogenes group.
  • Microbiological culture media
  • 500 g poly bottles