SP Wilmad-LabGlass Cylindrical Addition Funnels with Pressure Equalizing Sidearm, SP Industries
Supplier: SP Industries
LG-8266T-100
14207-458EA 536.2 USD
14207-458
SP Wilmad-LabGlass Cylindrical Addition Funnels with Pressure Equalizing Sidearm, SP Industries
Funnels Dropping Funnels
Featuring a PTFE stopcock, these borosilicate glass cylindrical funnels have a pressure equalizing sidearm
Standard Taper outer joint at the top is the same size as the lower Standard Taper inner joint. Inner joint has a sealed-in drip tube.