6-Mercapto-1-hexanol ≥97%
Supplier: Ambeed
|
Danger
|
A778998-25G
A778998-5G
A778998-1G
77627-544EA
8.24
USD
77627-544
77552-418
77552-416
6-Mercapto-1-hexanol ≥97%
6-Mercapto-1-hexanol
Caution: for Research Use Only
Formula:
C₆H₁₄OS MW: 134.24 g/mol |
MDL Number:
MFCD00068552 CAS Number: 1633-78-9 UN: 3334 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...