Dual Conductor Wrist Strap, 3M™, ORS Nasco
Supplier: ORS Nasco
051138-55935
12778-062EA 16.96 USD
12778-062
Dual Conductor Wrist Strap, 3M™, ORS Nasco
Consists of a blue, molded, insulative, thermoplastic exterior with an integrally molded conductive interior insert and low profile faceplate.
Lightweight and pliable thermoplastic material conforms to the arm to increase skin contact. 'Comfort bumps' on the interior insert help bring the band in contact with the skin, as well as allowing airflow between the band and skin for greater wearer comfort. The zipper-style latching mechanism easily adjusts to any size wrist. Band and housing are lot coded.