VWR® Advanced Hot Plate Stirrers
Supplier: VWR, part of Avantor
VWR®
984VW0CHSUSA 984VW4CHSUSA 984VW0AHSUSA 984VW7AHSUSA 984VW4AHSUSA 984VW7CHSUSA
97042-682EA 840.51 USD
97042-682 97042-602 97042-686 97042-646 97042-606 97042-642
VWR® Advanced Hot Plate Stirrers
Stirrers Magnetic Hotplate Stirrers
Advanced hot plate stirrers are designed to deliver accurate and repeatable results in general-purpose laboratory applications.
  • Excellent temperature uniformity
  • Industry leading safety features
  • Cool touch, chemical-resistant housing
  • LED display for temperature
  • Now with best-in-class, exclusive five-year warranty
These microprocessor controlled units feature a front-facing control panel with an Easy-to-read LED that displays set-point temperature.
Hot plate stirrers are available with ceramic or aluminum top plate material. Reflective white ceramic tops are chemical-resistant and easy to clean. Durable aluminum tops will not crack or chip, and offer a more even heating surface. A ramping feature that gradually increases speed results in improved safety, enhanced coupling, and excellent low end speed control. A built-in memory recall provides users with the last set temperature, even after the unit has been turned off. For additional safety, a hot symbol warning light is illuminated when the hot plate is in use, and remains lit until the top plate is sufficiently cooled.
Enhanced electronics regulate the robust heater, which brings samples to the desired temperature quickly and efficiently.Units are equipped with redundant safety controls, which will shut down unit in case of heater malfunction. Speed is precisely controlled, and provides consistent stirring at all speeds. The low-profile design utilizes minimal bench space and permits use within a fume hood. Unit housing is constructed of a heat- and chemical-resistant polymer that remains cool to the touch during operation. The spill-resistant design channels fluids away from internal components in the event of spills.
Certifications: RoHS certified. TUV listed. CE marked.
Ordering information: Units are supplied with a 234 cm (92") three-wire cord and plug. A 3.8 cm (11/2") PTFE coated stir bar is also included. The optional support rod and clamp kit includes a 45.7 cm (18") long stainless steel support rod, a thermometer/temperature probe extension clamp, a three-prong dual-adjust swivel clamp, and a hook connector. 230V, 50/60Hz models are also available; contact your VWR sales representative for more information.