SP Wilmad-LabGlass PTFE Stopcocks, with Metering Valve, SP Industries
Supplier: SP Industries
LG-9606T-102 LG-9606T-100
14204-418EA 180.14 USD
14204-418 14209-208
SP Wilmad-LabGlass PTFE Stopcocks, with Metering Valve, SP Industries
Straight, 1:5 taper PTFE stopcocks have a threaded needle valve
The needle valve extends through the bore of the plug and can be used as a valve within the stopcock to control flow. Stopcocks can be used for on-off control. Supplied complete with plug and O-ring set.