Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:
  • Search Within Results
You Searched for: "thymol-blue-sodium-salt-60-0-1-in-aqueous-solution"
SearchPresentationType-VERTICAL
SearchResultCount:"1"
Sort by:
Supplier:  Alfa Aesar
Description:   MDL: MFCD00151093 Notes: Transition interval: pH 1.2 (red) to pH 2.8 (yellow); pH 8.0 (yellow) to pH 9.2 (blue)