Would you like to log in again?
  • Criteria
    • SBE (2)
  • Supplier
  • Suppliers found in search results
    Sort by:
  • Search Within Results
You Searched for: "phenolphthalein-1-5-in-2-propanol"
SearchPresentationType-VERTICAL
SearchResultCount:"1"
Sort by:
Supplier:  Ricca Chemical
Description:   1 % (w/v) in 50% (v/v) Isopropanol. pH 8.0 (colorless) - 10.0 (pink). Neutralized - slight pink color is normal and desirable. Container: Plastic.
SDS
Small Business Enterprise