    Sort by:
  • Search Within Results
You Searched for: "bromocresol-green-0-1-60-1-in-aqueous-solution"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Ricca Chemical
Description:   0.1% (w/v) Aqueous Solution. pH 3.8 (yellow) - 5.4 (blue). ASTM, APHA for Acidity or Alkalinity. Plastic Container. 4L
SDS
Small Business Enterprise