Cleanroom Anti-Static General-Use Tape, Polypropylene, Ultratape
Supplier: UltraTape
2118CL200P5D
11212-050CS 1609.9 USD
11212-050
Cleanroom Anti-Static General-Use Tape, Polypropylene, Ultratape
Tapes
The nonstretch, antistatic polypropylene film features high antistatic performance with a surface resistivity of 109 ohms and a charge upon unwinding of no more than 100V
Ideal for general ESD use including sealing antistatic bags and boxes and static protection across sealed edges. Can also be used for packaging electronic parts and components, medical instruments, and precision mechanical equipment, and for film splicing. Contains no silicone. Plastic core is 7.6cm (3") in diameter.
Ordering information: Supplied double-bagged. For additional sizes, contact your VWR sales representative.