Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nalgene® 980 Braided Clear PVC Tubing, Thermo Scientific

Supplier: NALGE NUNC INTERNATIONAL
Nalgene®
8005-0360 8005-0260 8005-0040 8005-0130 8005-0070 8005-0190 8005-0100 8005-0310
Braided, reinforced PVC tubing is designed for high-pressure applications in the laboratory

Excellent for use with weak acids, weak bases, and salt solutions. Tubing has a high durometer hardness (Shore A, 65) that gives it extra flexibility, making it easier to bend into place and permitting tighter compression of crimp fittings for a secure fit. Complies with FDA regulations for food use. For traceability and easy reordering, the formulation number, I.D., and maximum working pressure are clearly imprinted in blue directly on the tubing.

Ordering information: Supplied in 50' coil lengths.



 

ORDER

Expand All / Collapse All
I.D. x O.D. Maximum Pressure Wall Thickness Supplier No. VWR Catalog Number Unit Price Quantity
1/2 x 3/4" 230 psig at 23°C (73°F), 115 psig at 52°C (125°F) 1/8" 8005-0190 63012-486 Pack of 50 Retrieving
1/4 x 7/16" 276 psig at 23°C (73°F), 138 psig at 52°C (125°F) 3/32" 8005-0070 63012-480 Pack of 50 Retrieving
3/16 x 3/8" 276 psig at 23°C (73°F), 138 psig at 52°C (125°F) 3/32" 8005-0040 63012-478 Pack of 50 Retrieving
3/4 x 1" 176 psig at 23°C (73°F), 88 psig at 52°C (125°F) 1/8" 8005-0310 63012-492 Pack of 50 Retrieving
3/8 x 9/16" 250 psig at 23°C (73°F), 125 psig at 52°C (125°F) 3/32" 8005-0130 63012-483 Pack of 50 Retrieving
5/16 x 1/2" 276 psig at 23°C (73°F), 138 psig at 52°C (125°F) 3/32" 8005-0100 63012-482 Pack of 50 Retrieving
5/8 x 7/8" 230 psig at 23°C (73°F), 115 psig at 52°C (125°F) 1/8" 8005-0260 63012-489 Pack of 50 Retrieving
1 x 15/16" 140 psig at 23°C (73°F), 70 psig at 52°C (125°F) 5/32" 8005-0360 63012-495 Pack of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty