Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nalgene® 180 Clear PVC Vacuum Tubing, Thermo Scientific

Supplier: NALGE NUNC INTERNATIONAL
Nalgene®
8000-1375 8000-0055 8000-1210 8000-0065 8000-0145 8000-0210 8000-1145 8000-1065 8000-1270 8000-1325
Clear PVC tubing has heavy walls to withstand higher working pressure

Durometer hardness: Shore A, 55. Autoclavable. Complies with USP Class VI, Food-Grade and USDA standards.

Ordering information: Supplied in 10' or 50' coil lengths.



 

ORDER

Expand All / Collapse All
Coil Length I.D. x O.D. Maximum Pressure Wall Thickness Supplier No. VWR Catalog Number Unit Price Quantity
15.24 m (50') 1/4 x 5/8" 56 psig @ 23°C (73°F), 28 psig @ 52°C (125°F) 3/16" 8000-0065 63013-404 Pack of 50 Retrieving
15.24 m (50') 3/16 x 9/16" 62 psig @ 23°C (73°F), 31 psig @ 52°C (125°F) 3/16" 8000-0055 63013-492 Pack of 50 Retrieving
15.24 m (50') 3/8 x 7/8" 54 psig @ 23°C (73°F), 27 psig @ 52°C (125°F) 1/4" 8000-0145 63013-437 Pack of 50 Retrieving
15.24 m (50') 1/2 x 11/8" 52 psig @ 23°C (73°F), 26 psig @ 52°C (125°F) 5/16" 8000-0210 63013-517 Pack of 50 Retrieving
3.05 m (10') 1/2 x 11/8" 52 psig @ 23°C (73°F), 26 psig @ 52°C (125°F) 5/16" 8000-1210 63013-787 Case (10Feet) Retrieving
3.05 m (10') 1/4 x 5/8" 56 psig @ 23°C (73°F), 28 psig @ 52°C (125°F) 3/16" 8000-1065 63013-763 Case (10Feet) Retrieving
3.05 m (10') 3/8 x 7/8" 54 psig @ 23°C (73°F), 27 psig @ 52°C (125°F) 1/4" 8000-1145 63013-783 Case (10Feet) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty