Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nalgene® 180 Clear PVC Tubing, Thermo Scientific

Supplier: NALGE NUNC INTERNATIONAL
Nalgene®
8000-0110 8000-0010 8000-0310 8000-0350 8000-0130 8000-0030 8000-9120 8000-9020 8000-5120 8000-0190 8000-0090 8000-0070 8000-9180 8000-5060 8000-4090 8000-9060 8000-0120 8000-0200 8000-0100 8000-0260 8000-4200 8000-0160 8000-0020 8000-4120 8000-0140 8000-1350 8000-0006 8000-9030 8000-9010 8000-0080 8000-4140 8000-0060 8000-9090 8000-0180 8000-4060 8000-1090 8000-4080
A high-quality, lab-grade PVC tubing that is both economical and versatile

All coils are marked continuously with I.D. and regulatory compliance. Also marked at one-foot intervals for convenient measuring and cutting. Autoclavable. Durometer hardness: Shore A, 55.

Flexible tubing is made from the highest grade resins and special plasticizers; no fillers or extenders added. Resists a wide range of chemicals. Dimensionally stable and highly resistant to hardening and discoloration.

Certifications: Complies with the Food Additives Amendment of the Federal Food, Drug, and Cosmetic Act and with U.S. Pharmacopoeia Class VI Requirements for Plastic Materials.

Ordering information: Lengths of 10', 50', 100' and 250' come in a specially designed box for cleanliness, easy dispensing, and storage. 500' lengths available on reels.



 

ORDER

Expand All / Collapse All
Coil Length I.D. x O.D. Maximum Pressure Wall Thickness Supplier No. VWR Catalog Number Unit Price Quantity
10' 5/16 x 7/16" 30 psig @ 23°C, 15 psig @ 52°C 1/16" 8000-1090 63013-773 Case of 5 (10 Feet) Retrieving
Pack of 1 (10Feet) Retrieving
100' 1/2 x 3/4" 34 psig @ 23°C, 17 psig @ 52°C 1/8" 8000-4200 63013-820 Case (100Feet) Retrieving
100' 3/8 x 5/8" 44 psig @ 23°C, 22 psig @ 52°C 1/8" 8000-4140 63013-816 Case (100Feet) Retrieving
100' 1/4 x 3/8" 34 psig @ 23°C, 17 psig @ 52°C 1/16" 8000-4060 63013-808 Case (100Feet) Retrieving
100' 1/4 x 1/2" 52 psig @ 23°C, 26 psig @ 52°C 1/8" 8000-4080 63013-810 Case (100Feet) Retrieving
100' 3/8 x 1/2" 25 psig @ 23°C, 13 psig @ 52°C 1/16" 8000-4120 63013-814 Case (100Feet) Retrieving
250' 5/16 x 7/16" 30 psig @ 23°C, 15 psig @ 52°C 1/16" 8000-9090 63013-685 Case of 250 Retrieving
250' 1/8 x 1/4" 50 psig @ 23°C, 25 psig @ 52°C 1/16" 8000-9020 63013-608 Case of 1 Retrieving
250' 3/16 x 5/16" 40 psig @ 23°C, 20 psig @ 52°C 1/16" 8000-9030 63013-620 Case of 250 Retrieving
250' 1/2 x 5/8" 22 psig @ 23°C, 11 psig @ 52°C 1/16" 8000-9180 63013-732 Case of 250 Retrieving
50' 1 x 11/4" 22 psig @ 23°C, 11 psig @ 52°C 1/8" 8000-0350 63013-211 Pack of 50 Retrieving
50' 1/4 x 1/2" 52 psig @ 23°C, 26 psig @ 52°C 1/8" 8000-0080 63013-415 Pack of 50 Retrieving
50' 1/4 x 3/8" 34 psig @ 23°C, 17 psig @ 52°C 1/16" 8000-0060 63013-244 Pack of 50 Retrieving
50' 5/16 x 7/16" 30 psig @ 23°C, 15 psig @ 52°C 1/16" 8000-0090 63013-426 Pack of 50 Retrieving
50' 3/16 x 5/16" 40 psig @ 23°C, 20 psig @ 52°C 1/16" 8000-0030 63013-379 Pack of 50 Retrieving
50' 5/16 x 9/16" 46 psig @ 23°C, 23 psig @ 52°C 1/8" 8000-0110 63013-390 Pack of 50 Retrieving
50' 3/8 x 5/8" 44 psig @ 23°C, 22 psig @ 52°C 1/8" 8000-0140 63013-197 Pack of 50 Retrieving
50' 1/8 x 3/16" 34 psig @ 23°C, 17 psig @ 52°C 1/32" 8000-0010 63013-357 Pack of 50 Retrieving
50' 1/2 x 5/8" 22 psig @ 23°C, 11 psig @ 52°C 1/16" 8000-0180 63013-233 Pack of 50 Retrieving
50' 5/16 x 1/2" 40 psig @ 23°C, 20 psig @ 52°C 3/32" 8000-0100 63013-506 Pack of 50 Retrieving
50' 3/4 x 1" 26 psig @ 23°C, 13 psig @ 52°C 1/8" 8000-0310 63013-164 Pack of 50 Retrieving
50' 1/2 x 11/16" 28 psig @ 23°C, 14 psig @ 52°C 3/32" 8000-0190 63013-255 Pack of 50 Retrieving
50' 7/16 x 5/8" 34 psig @ 23°C, 17 psig @ 52°C 3/32" 8000-0160 63013-277 Pack of 50 Retrieving
50' 3/8 x 1/2" 26 psig @ 23°C, 13 psig @ 52°C 1/16" 8000-0120 63013-288 Each (50Feet) Retrieving
50' 1/2 x 3/4" 34 psig @ 23°C, 17 psig @ 52°C 1/8" 8000-0200 63013-448 Pack of 50 Retrieving
50' 3/8 x 9/16" 36 psig @ 23°C, 18 psig @ 52°C 3/32" 8000-0130 63013-175 Pack of 50 Retrieving
50' 1/4 x 7/16" 44 psig @ 23°C, 22 psig @ 52°C 3/32" 8000-0070 63013-222 Pack of 50 Retrieving
50' 1/8 x 1/4" 50 psig @ 23°C, 25 psig @ 52°C 1/16" 8000-0020 63013-368 Pack of 50 Retrieving
50' 5/8 x 7/8" 28 psig @ 23°C, 14 psig @ 52°C 1/8" 8000-0260 63013-470 Pack of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty