Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR Signature™ Clear PVC Tubing

Supplier: VWR International
VWR® Signature™
408000-0322 408000-0200 408000-0120 408000-0030 408000-0140 408000-0350 408000-0020 408000-0070 408000-0060 408000-0090 408000-0080
VWR Signature™ Clear PVC Tubing is engineered for optimum performance in a wide variety of applications.

  • Lab-grade clear plastic tubing
  • Wide operating temperature range
  • Autoclavable

Designed for use in medical research, clinical, pharmaceutical, food and beverage, media manufacture and transfer, and vapor transfer applications

Durometer hardness: Shore A, 55. Operating temperature range: -32 to 71°C (-25 to 160°F). Tensile strength: 1650 psi. Complies with FDA, USP Class VI and USDA standards.

Can also be used with peristaltic pumps, CO2 incubators, and condenser coolant lines. This autoclavable, lab-grade clear plastic tubing is dimensionally stable and highly resistant to hardening and discoloration. Made from the highest grade resins and special plasticizers (no fillers or extenders are added). Resists a wide range of chemicals.

Ordering information: Supplied in 50' coil lengths.



 

ORDER

Expand All / Collapse All
I.D. x O.D. Wall Thickness VWR Catalog Number Unit Price Quantity
1/2 x 3/4" 1/8" 60985-556 Each (50Feet) Retrieving
1/4 x 1/2" 1/8" 60985-532 Each (50Feet) Retrieving
1/4 x 3/8" 1/16" 60985-528 Each (50Feet) Retrieving
1/4 x 7/16" 3/32" 60985-530 Each (50Feet) Retrieving
1/8 x 1/4" 1/16" 60985-512 Each (50Feet) Retrieving
3/4 x 11/4" 1/4" 60985-578 Each (50Feet) Retrieving
3/8 x 1/2" 1/16" 60985-540 Each (50Feet) Retrieving
3/8 x 5/8" 1/8" 60985-544 Each (50Feet) Retrieving
5/16 x 7/16" 1/16" 60985-534 Each (50Feet) Retrieving
1 x 11/4" 1/8" 60985-584 Each (50Feet) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty