Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Histoplex™ Tissue Processing Cassettes with Micromesh Chamber, Starplex®

Supplier: STARPLEX SCIENTIFIC INC
Histoplex™
TC-ORANGE TC-BLUE TC-AQUA TC-PEACH TC-GREY TC-DRKGREEN TC-PINK TC-LILAC TC-RED TC-WHITE TC-GREEN TC-YELLOW TC-TAN
Ideal for embedding, processing, and storage of standard tissue specimens as well as tissue fragments

Cassettes are designed with an innovative micromesh chamber to securely retain fragments, leaving substantial room for additional, larger specimens. This 0.23mm micromesh chamber provides effective fluid exchange and eliminates the need for additional filters. The durable acetyl polymer construction is resistant to the chemical action of histology agents. Cassettes snap open or shut easily and feature a 45° angled writing surface that is suitable for use with most labeling devices. Available in 13 non-metallic colors. Non-cytotoxic.

Ordering information: Cassettes are packaged 500 per dispenser box, 3 boxes per case.



 

ORDER

Expand All / Collapse All
Color Supplier No. VWR Catalog Number Unit Price Quantity
Aqua TC-AQUA 15001-166 Case of 1,500 Retrieving Restricted
Blue TC-BLUE 15001-140 Case of 1,500 Retrieving Restricted
Dark Green TC-DRKGREEN 15001-158 Case of 1,500 Retrieving Restricted
Gray TC-GREY 15001-156 Case of 1,500 Retrieving Restricted
Green TC-GREEN 15001-148 Case of 1,500 Retrieving Restricted
Lilac TC-LILAC 15001-154 Case of 1,500 Retrieving Restricted
Orange TC-ORANGE 15001-146 Case of 1,500 Retrieving Restricted
Peach TC-PEACH 15001-160 Case of 1,500 Retrieving Restricted
Pink TC-PINK 15001-144 Case of 1,500 Retrieving Restricted
Red TC-RED 15001-152 Case of 1,500 Retrieving Restricted
Tan TC-TAN 15001-150 Case of 1,500 Retrieving Restricted
White TC-WHITE 15001-138 Case of 1,500 Retrieving Restricted
Yellow TC-YELLOW 15001-142 Case of 1,500 Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty