Would you like to log in again?
SHURMARK® PLUS Color-Frosted Microscope Slides, Triangle Biomedical
Supplier: General Data Healthcare
SHUR/Mark®
SMP-STT-45W SM-PCTS-G SMP-STT-45Y SMP-SC-TT-45P SMP-SCTT-45L SMP-SCTT-B SMP-SCTT-45G SMP-STT-B SMP-STT-BG SMP-STT-BB SMP-STT-G SMP-STT-BP SMP-STT-L SMP-SCTT-45W SMP-SCTT-45Y SMP-STT-BL SMP-STT-P SMP-SCTT-BP SMP-STT-BW SMP-SCTT-W SMP-SCTT-BL SMP-SCTT-Y SMP-STT-W SMP-SCTT-BY SMP-STT-Y SMP-STT-BY SMP-SCTT-BW SMP-STT-45G SMP-SCTT-BB SMP-SCTT-L SMP-SCTT-G SMP-STT-45B SMP-STT-45P SMP-SCTT-BG SMP-SCTT-P SMP-STT-45L
SHURMark PLUS Color-Frosted Microscope Slides are compatible with Leica and Thermo Slide Labelers
Choice of processing or biopsy cassette taped sleeves. Both types are specifically designed and quality controlled for use with the SHURMark® PLUS cassette instrument.
They have ground and polished edges and are available in six popular colors. Thery are compatible with Ink-Jet labelers and can be easily be written on with pen or pencil.
ORDER
Expand All / Collapse All
Description L x W Thickness Color Supplier No. VWR Catalog Number Unit Price Quantity
Charged with 45° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Yellow SMP-SCTT-45Y 89413-518 Case of 1,440 Retrieving
Charged with 45° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Blue SMP-SCTT-BB 89413-536 Case of 1,440 Retrieving
Charged with 45° Clipped Corner 75 x 25 mm (3 x 1") 1 mm White SMP-SCTT-45W 89413-510 Case of 1,440 Retrieving
Charged with 45° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Lilac SMP-SCTT-45L 89413-520 Case of 1,440 Retrieving
Charged with 45° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Green SMP-SCTT-45G 89413-514 Case of 1,440 Retrieving
Charged with 45° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Pink SMP-SC-TT-45P 89413-516 Case of 1,440 Retrieving
Charged with 90° Clipped Corner 75 x 25 mm (3 x 1") 1 mm White SMP-SCTT-W 10010-880 Case of 1440 Retrieving Restricted
Pack of 72 Retrieving Restricted
Small Business Enterprise
Charged with 90° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Yellow SMP-SCTT-Y 10010-890 Case of 1440 Retrieving Restricted
Pack of 72 Retrieving Restricted
Small Business Enterprise
Charged with 90° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Blue SMP-SCTT-B 10010-882 Case of 1440 Retrieving Restricted
Pack of 72 Retrieving Restricted
Small Business Enterprise
Charged with 90° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Lilac SMP-SCTT-L 10010-896 Case of 1440 Retrieving Restricted
Pack of 72 Retrieving Restricted
Small Business Enterprise
Charged with 90° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Pink SMP-SCTT-P 10010-886 Case of 1440 Retrieving Restricted
Pack of 72 Retrieving Restricted
Small Business Enterprise
Charged with 90° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Green SMP-SCTT-G 10010-884 Case of 1440 Retrieving Restricted
Pack of 72 Retrieving Restricted
Small Business Enterprise
Charged with Beveled Corner 75 x 25 mm (3 x 1") 1 mm Green SMP-SCTT-BG 89413-538 Case of 1,440 Retrieving
Charged with Beveled Corner 75 x 25 mm (3 x 1") 1 mm Pink SMP-SCTT-BP 89413-540 Case of 1,440 Retrieving
Charged with Beveled Corner 75 x 25 mm (3 x 1") 1 mm Yellow SMP-SCTT-BY 89413-542 Case of 1,440 Retrieving
Charged with Beveled Corner 75 x 25 mm (3 x 1") 1 mm Lilac SMP-SCTT-BL 89413-544 Case of 1,440 Retrieving
UnCharged with 45° Clipped Corner 75 x 25 mm (3 x 1") 1 mm White SMP-STT-45W 89413-498 Case of 1,440 Retrieving
UnCharged with 45° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Blue SMP-STT-45B 89413-500 Case of 1,440 Retrieving
UnCharged with 45° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Green SMP-STT-45G 89413-502 Case of 1,440 Retrieving
UnCharged with 45° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Pink SMP-STT-45P 89413-504 Case of 1,440 Retrieving
UnCharged with 45° Clipped Corner 75 x 25 mm (3 x 1") 1 mm White SMP-STT-45W 10029-564 Case of 20 Retrieving Restricted
UnCharged with 45° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Yellow SMP-STT-45Y 89413-506 Case of 1,440 Retrieving
UnCharged with 45° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Lilac SMP-STT-45L 89413-508 Case of 1,440 Retrieving
UnCharged with 90° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Lilac SMP-STT-L 10010-878 Case of 1440 Retrieving Restricted
Pack of 72 Retrieving Restricted
Small Business Enterprise
UnCharged with 90° Clipped Corner 75 x 25 mm (3 x 1") 1 mm White SMP-STT-W 10010-862 Case of 1440 Retrieving Restricted
Pack of 72 Retrieving Restricted
Small Business Enterprise
UnCharged with 90° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Pink SMP-STT-P 10010-868 Case of 1440 Retrieving Restricted
Pack of 72 Retrieving Restricted
Small Business Enterprise
UnCharged with 90° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Blue SMP-STT-B 10010-864 Case of 1440 Retrieving Restricted
Pack of 72 Retrieving Restricted
Small Business Enterprise
UnCharged with 90° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Yellow SMP-STT-Y 10010-872 Case of 1440 Retrieving Restricted
Pack of 72 Retrieving Restricted
Small Business Enterprise
UnCharged with 90° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Green SMP-STT-G 10010-866 Case of 1440 Retrieving Restricted
Pack of 72 Retrieving Restricted
Small Business Enterprise
UnCharged with Beveled Corner 75 x 25 mm (3 x 1") 1 mm Green SMP-STT-BG 89413-526 Case of 1,440 Retrieving
UnCharged with Beveled Corner 75 x 25 mm (3 x 1") 1 mm Pink SMP-STT-BP 89413-528 Case of 1,440 Retrieving
UnCharged with Beveled Corner 75 x 25 mm (3 x 1") 1 mm Lilac SMP-STT-BL 89413-532 Case of 1,440 Retrieving
UnCharged with Beveled Corner 75 x 25 mm (3 x 1") 1 mm Yellow SMP-STT-BY 89413-530 Case of 1,440 Retrieving
UnCharged with Beveled Corner 75 x 25 mm (3 x 1") 1 mm Blue SMP-STT-BB 89413-524 Case of 1,440 Retrieving
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty