VWR® UrineTime II™ Proportioning Devices
Supplier: VWR, part of Avantor
4010-420-310
93001-122CS 216.76 USD
93001-122
VWR® UrineTime II™ Proportioning Devices
Ideal for outpatient use, this 24-hour urine collection proportioning system is designed for the automatic division of samples into two separate containers for chemistry assays.
  • Supplied with a plastic bag
  • Designed for outpatient use
  • 24-hour urine collection system
  • Automatically divides samples into separate containers for analysis
Useful for analysis requiring two separate preparations or preservatives. The two 2L bottles feature keyed sides that stay firmly locked together. A plastic bag is supplied for safe disposal of the removable plastic funnel.