Noc-5, Calbiochem®

Synonyms: 1-Hydroxy-2-oxo-3-(3-aminopropyl)-3-isopropyl-1-triazene

487950-10MG
CA80053-446
Noc-5, Calbiochem®
Noc-5

A nitric oxide (NO) donor. A stable NO-amine complex that can release two equivalents of NO spontaneously in solution under physiological conditions without any cofactor. Half-life (t1/2) of NO release=93min. (PBS, pH 7.4, 22°C, 72°F).

Formula: C₆H₁₆N₄O₂
CAS Number: 146724-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR