Demecolcine, Calbiochem®

Synonyms: Colcemid

234109-5MG 234109-5MG
80055-172EA 128.99 USD
CA80055-172 80055-172
Demecolcine, Calbiochem®
Demecolcine

Cell synchronization agent


Induces apoptosis by blocking mitosis in HeLa S3 cells. Depolymerizes microtubules and limits microtubule formation. Appropriate dilutions inactivate spindle fiber mechanism during metaphase, systematically immobilizing chromosomes at the metaphase plate.

Formula: C₂₁H₂₅NO₅
MW: 371.433 g/mol
MDL Number: MFCD00075459
CAS Number: 477-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR