Etoposide, Calbiochem®

Supplier: MilliporeSigma

Synonyms: (5R,5aR,8aR,9S)-9-(((4aR,6R,7R,8R,8aS)-7,8-Dihydroxy-2-methylhexahydropyrano[3,2-d][1,3]dioxin-6-yl)oxy)-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,5a,8a,9-tetrahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(8H)-one4'-Demethylepipodophyllotoxin 9-(4,6-O-Ethylidene-beta-D-glucopyranoside)

341205-25MG 341205-25MG
80055-248EA 103.56 USD
80055-248 CA80055-248
Etoposide, Calbiochem®
Etoposide

Topoisomerase II inhibitor (IC50=59.2µM). A derivative of podophyllotoxin that has major activity against a number of tumors, including germ cell neoplasms, small cell lung cancer, and malignant lymphoma.

Formula: C₂₉H₃₂O₁₃
MW: 588.56 g/mol
MDL Number: MFCD00869325
CAS Number: 33419-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR