LY-83583, Calbiochem®

Supplier: MilliporeSigma

Synonyms: 6-Anilinoquinoline-5,8-quinone

440205-5MG 440205-5MG
80056-982EA 120.69 USD
80056-982 CA80056-982
LY-83583, Calbiochem®
LY-83583

A competitive inhibitor of soluble guanylate cyclase (IC50=2µM). Lowers the production of cGMP levels in a wide range of tissues by blocking intracellular Ca2+ release, with negligible effect on cAMP levels. Inhibits nitric oxide-induced smooth muscle relaxation. Shown to inhibit interleukin-1-induced cGMP accumulation in cultured rat aortic vascular smooth muscle cells.

Formula: C₁₅H₁₀N₂O₂
MW: 250.25 g/mol
CAS Number: 91300-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR