SIN-1 chloride, Calbiochem®

Supplier: MilliporeSigma

Synonyms: Sin-1 Hydrochloride

567028-20MG 567028-20MG
80054-462EA 162.66 USD
CA80054-462 80054-462
SIN-1 chloride, Calbiochem®
SIN-1 chloride

Nitric oxide (NO) donor. Spontaneously decomposes to yield NO and superoxide anion radicals. Also inhibits the release of plasminogen activator inhibitor (PAI) from stimulated platelets.

Formula: C₆H₁₁ClN₄O₂
CAS Number: 16142-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR