Shoe brush machine has an automatic sensor unit that is activated when foot is placed in the opening.
- 99.99% effective HEPA filter exhaust system prevents particles from re-entering the surrounding area
- Captures heavy contaminants in a convenient pull-out drawer
- Self-contained machine includes a built-in vacuum system and a timing belt drive between the gear motor and bottom brush assembly
Operating noise of blower, vacuum, and gear motor is less than 55 dBA.
Delivery information: Shoe brush machine includes an easily removable dust drawer, HEPA filter, and vacuum bag, a rotomolded polyethylene cover, and four motor driven brushes.