This shoe brush machine is designed with four transmission-driven brushes, including a floating weight-loaded top brush and rotating bottom brush that clean sides, top, and bottom of shoes prior to entry into contamination-controlled areas.
Rubber shin guard and cast iron base
Safety control switch and instructions are located on the support handle to assist in maintaining balance
white polyethylene cover with a removable hood secured by four latches
For 110 V, 60 Hz. 1/3 hp motor
Dimension: 51 x 47 x 99 cm (20 x 18 1/2x 39")
Certifications: Complies with U.S. Air Force Tech. Order T.O. 00-25-203 and Federal Standard 209.
Ordering information: Machine should be used with a vacuum system, sold separately. Unit dimensions are given with support handle included.