Calbiochem® Caspase-1 Inhibitor IV
Supplier: EMD Millipore
Calbiochem®
400015-1MG 400015-1MG
80052-274EA 161.5 USD
80052-274 CA80052-274
Calbiochem® Caspase-1 Inhibitor IV
Proteins and Peptides
A highly selective, competitive, and irreversible inhibitor of caspase-1 and caspase-4
Inactivates the enzyme with a rate limited by diffusion and is relatively inert toward other bionucleophiles such as glutathione, making it an excellent candidate for in vivo studies of enzyme inhibition.