Calbiochem® Group III Caspase Inhibitor I, EMD Millipore
Supplier: EMD Millipore
Calbiochem®
368620-1MG 368620-1MG
80059-152EA 482.56 USD
80059-152 CA80059-152
Calbiochem® Group III Caspase Inhibitor I, EMD Millipore
Proteins and Peptides
A potent, cell-permeable, and irreversible inhibitor of Group III caspases (caspase-6, -8, -9, and -10) although more effective towards caspases-6 and -8.