Would you like to log in again?
Polyblend Boots, Onguard
Supplier: ONGUARD INDUSTRIES
86102-13 86102-12 86102-11 86102-9 86102-10
PU/PVC boots feature two-stage, injection-molded, one-piece construction for complete waterproof protection.
  • Protective steel toe cap
  • Non-slip soles give workers positive footing
  • Pull-up/kick-off lugs make entry and removal easy
  • Steel shank adds comfort
  • Removable cushion insole permits cleaning of boot interior.
  • Choose plain toe and steel toe styles in five sizes
Polyurethane/PVC composition provides a high degree of chemical resistance.
Color: Black
Certifications: CSA Z195 M92 Grade 1 certified. Steel toe series meets or exceeds ANSI Z41 PT91 I/75 C/75 standard for impact and compression.
ORDER
Expand All / Collapse All
Shoe Size Color Size Supplier No. VWR Catalog Number Unit Price Quantity
10 Black 10 86102-10 21921-736 Pack of 1 Retrieving
Pair Retrieving
11 Black 11 86102-11 21921-738 Pack of 1 Retrieving
12 Black 12 86102-12 21921-740 Pack of 1 Retrieving
13 Black 13 86102-13 21921-742 Pack of 1 Retrieving
9 Black 9 86102-9 21921-734 Pack of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty