Human Recombinant Caspase-10 (from E. coli)
Supplier: MilliporeSigma
218785-3000U 218785-3000U
80059-010EA 601.92 USD
CA80059-010 80059-010
Human Recombinant Caspase-10 (from E. coli)
Proteins and Peptides
In 100mM NaCl, 50mM HEPES, 10mM DTT, 1mM EDTA, 10% glycerol, 0.1% CHAPS, pH 7.4
Useful for the study of enzyme regulation, kinetics, cleavage of target substrates, and inhibitor screening. Activity: >=50,000units/mL. Specific activity: >=150,000units/mg protein. U is equal to 1 unit.