Human Recombinant NADPH-P450 Oxidoreductase
Supplier: EMD Millipore
481974-200UG 481974-200UG
80053-352EA 736.16 USD
80053-352 CA80053-352
Human Recombinant NADPH-P450 Oxidoreductase
In 10 mM potassium phosphate buffer, 1 mM DTT, 100 µM EDTA, 20% glycerol, pH 7.7
A membrane bound flavoprotein containing one mole each of FAD and FMN. Transfers electrons from NADPH to the various isoforms of cytochrome P450. Typically used at ratios ranging from 1:10 to 1:2 (reductase: P450) for in vitro reconstitution assays. Suitable for in vitro drug metabolism and toxicology research including reconstituted P450-dependent biotransformation reactions. Specific activity: >=20 units/mg protein.