Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Empore™ Extraction Disks, 3M™

Supplier: 3M BIOANALYTICAL TECHNOLOGIES
Empore®
FA400 2252 2241 2340 2240 2272 2370 2270 2315 2215 2214 98-0405-0062-5 98-0405-0063-3
Disks are thin, particle-loaded membranes, with the particles embedded in a stable, inert matrix of PTFE fibrils

Disks eliminate emulsion formation and provide excellent reproducibility. Can be used for extracting semi-volatile organics, polar compounds, ionic compounds, explosives, and pesticides.

Approved for use with numerous EPA methods, including 525.2 and 1664 for oil and grease detection.

Ideal for achieving faster extractions with lower solvent usage and better sample concentrations.



 

ORDER

Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Disks, C8, 47 mm 2214 55004-104 Case of 60 Retrieving
Pack of 20 Retrieving
Disks, C18, 47 mm 2215 55004-098 Case of 60 Retrieving
Pack of 20 Retrieving
Disks, SDB-XC, 47 mm 2240 55004-086 Case of 60 Retrieving
Pack of 20 Retrieving
Disks, C18, 90 mm 2315 55004-096 Case of 30 Retrieving
Pack of 10 Retrieving
Disks, SDB-XC, 90 mm 2340 55004-084 Case of 30 Retrieving
Pack of 10 Retrieving
Filter Aid 400 FA400 55004-048 Case (1,500g) Retrieving
Oil and Grease Disks, C18, 47 mm 98-0405-0062-5 76008-564 Case of 3 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty