Resprep® SPE Cartridges, Restek
Supplier: Restek
Resprep®
26063
RK26063PK 1119.49 USD
RK26063
Resprep® SPE Cartridges, Restek
Chromatography Columns
Resprep® SPE cartridges contain hydrophilic adsorbents used to extract hydrophilic analytes from nonpolar matrices, such as organic solvents.
  • Constructed of PTFE frits or glass tubes with PTFE frits
EPA method 548.1 is specifically designed to provide consistent and reproducible results for the extraction of endothal from aqueous samples. Weak anion exchange resin packing (BioRex 5).
Packaging: Packed with weak anion exchange resin (BioRex 5).