Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Accessories

Supplier: HAMILTON COMPANY
14806




 

ORDER

Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Syringe Guide for 5–10 µL 700 and 1700 Series and Models 75, 701 Syringes 14806 60373-425 Each Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
For Tubes Description For Use With O.D. Needle Gauge Size Supplier No. VWR Catalog Number Unit Price Quantity
- 700, 1700 and 7000 Series Syringes - - - - 204880 89168-730 Each Retrieving Restricted
Cleaning Concentrate, 500 mL - - - - - 18311 60380-806 Each Retrieving Restricted
Cleaning Concentrate, 70 mL - - - - - 18310 60380-781 Each Retrieving Restricted
Hamilton Repeating Dispenser for 25 µL - 2.5 mL Volume Syringes - Models 701, 702, 705, 710, 725, and 750 - - - 83700 60384-002 Each Retrieving Restricted
Needle Cleaning Kit with Tungsten Cleaning Wires and 70 mL Bottle of Cleaning Concentrate - - 76620A 60380-759 Each Retrieving Restricted
Reproducibility (Chaney) Adapter for 1702–1750 Series, 702–750 Series, or 7000 Series Syringes - - - - - 14725 60374-016 Each Retrieving Restricted
Syringe Guide for 7000 Series and 25–500 µL 700 and 1700 Series Syringes - - - - - 14906 60374-018 Each Retrieving Restricted
Tungsten Needle Cleaning Wires - - 0.306 mm 22, 23, and Larger 17.8 cm 18304 72310-062 Pack of 10 Retrieving Restricted
Tungsten Needle Cleaning Wires - - 0.207 mm 24–26 17.8 cm 18303 72310-064 Pack of 10 Retrieving Restricted
Tungsten Needle Cleaning Wires - - 0.167 mm 27 17.8 cm 18302 72310-066 Pack of 10 Retrieving Restricted
Tungsten Needle Cleaning Wires - - 0.126 mm 22S, 25S, and 28–30 17.8 cm 18301 72310-068 Pack of 10 Retrieving Restricted
Tungsten Needle Cleaning Wires - - 0.089 mm 26S and 31–33 17.8 cm 18300 72310-070 Pack of 10 Retrieving Restricted
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty