SP Wilmad-LabGlass Flash Chromatography Flow Controller, SP Industries
Supplier: SP Industries
LG-4520-100
14209-544EA 352.31 USD
14209-544
SP Wilmad-LabGlass Flash Chromatography Flow Controller, SP Industries
Flow Controllers
These flow controllers control pressure applied to a reservoir or directly to a chromatography column
  • Borosilicate glass
  • For analytical and preparative separations
  • PTFE valve and vent
They are made of borosilicate glass and are designed to affect rapid separations in both analytical and preparative modes. The PTFE valve and vent control the pressure to reservoir/column. Top valve is 0 to 4 mm.
Ordering information: Complete assembly consists of column, reservoir, flow controller, and clamps. See 14210-628 series for reservoirs and 14208-788 series for columns.