Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

556 MPS Multiparameter Logging System, YSI

Supplier: XYLEM/YSI
5563-4 5563-10 5563-20 556-01 556-02 005560
This handheld IP67 waterproof system simultaneously measures dissolved oxygen (DO), conductivity, temperature, salinity, total dissolved solids, and with optional probe, pH and ORP

Membrane kits include six polyethylene membranes and one bottle of KCl solution. Membranes have decreased stirring requirements and fast response times. A flow cell is available for ground water applications.

IP67 rating means that the meter can be immersed in water to a depth of 1 m for 1 hour without any water incursion. Stores up to 49,000 sets of time/date stamped data. Features easy-to-use, screw-on cap DO membranes, field-replaceable probes, and an RS 232 interface. An optional internal barometer can be user-calibrated and displayed along with other data, used in DO calibrations, and logged to memory for tracking changes in barometric pressure. System features on-screen parameter graphing and stores calibration data to assist in Good Laboratory Practices (GLP).

Ordering information: Three-year manufacturer's warranty on the instrument; one-year manufacturer's warranty on the probe module and cables. Instrument comes with a handstrap, instruction manual, four C-size alkaline batteries, PC interface cable, membrane kit (14236-070), and a large soft-sided carrying case. Large case can hold the instrument, a 20 m (65'7") cable, and additional calibrating supplies. Form-fitted carrying case contains the instrument and features a shoulder strap. Small carrying cases 13501-990 and -962 fit the instrument and a 4 m (13'1") cable.



 

ORDER

Expand All / Collapse All

DO/Temperature/Conductivity Probes
Description Supplier No. VWR Catalog Number Unit Price Quantity
Probe with 4 m (13'1") Cable 5563-4 13501-966 Each Retrieving Restricted
Probe with 10 m (32'7") Cable 5563-10 13501-968 Each Retrieving Restricted
Probe with 20 m (65'7") Cable 5563-20 13501-972 Each Retrieving Restricted
Probe without cable 005560 10218-816 Each (1lb) Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Dimensions
Pressure range
Temperature Range
Weight
Temperature Accuracy
Temperature Resolution
Pressure Resolution
Pressure Accuracy
pH Range
pH Resolution
pH accuracy
Conductivity Range
Conductivity Resolution
Conductivity Accuracy
Salinity Range
Salinity Resolution
Salinity Accuracy
DO Accuracy
ORP resolution
ORP accuracy
DO Range
DO Resolution
ORP range
Total Dissolved Solids Range
Total Dissolved Solids Resolution

ACCESSORIES

Expand All / Collapse All

Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
Cigarette Lighter Charger 063516 13501-986 Each Retrieving Restricted
Flow Cell 005083 13501-970 Each Retrieving Restricted
Form-Fitted Carrying Case, with Shoulder Strap 655259 13501-982 Each Retrieving Restricted
Hands-Free Harness 655227 13501-984 Each Retrieving Restricted
Membrane Kit 605306 14236-068 Pack of 6 Retrieving Restricted
Membrane Kit, 2.00 mil 605307 14236-070 Pack of 6 Retrieving Restricted
pH/ORP Probe 05565 13501-974 Each Retrieving Restricted
pH Probe 005564 13501-976 Each Retrieving Restricted
Rechargeable Battery Kit 655277 13501-988 Each Retrieving Restricted
Replacement DO Electrode 655900 13500-922 Each Retrieving Restricted
Small Soft-Sided Carrying Case 655100 13501-990 Each Retrieving Restricted
Ultra Clamp C-Clamp Mount 063517 13501-980 Each Retrieving Restricted
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty