Sodium butyrate ≥98%, Calbiochem®
Supplier: MilliporeSigma
Synonyms:
Butyric acid sodium salt
, Sodium Butyrate
567430-250MG
567430-250MG
80054-470EA
99.77
USD
CA80054-470
80054-470
Sodium butyrate ≥98%, Calbiochem®
Sodium butyrate
A physiologically produced short chain fatty acid that has been shown to induce apoptosis in colon carcinoma cell lines by a p53-independent pathway. Inhibits c-myc splicing and interferes with signal transduction processes, including the release of Ca2+ from intracellular stores. Reduces histamine-induced calcium mobilization in C6 glioma cells. Down-regulates the activity of the serine/threonine protein kinase casein kinase II in HT29 cells.
Formula:
C₄H₇NaO₂ MW: 110.09 g/mol Melting Pt: 244 °C Density: 1.324 g/cm³ (30 °C) |
MDL Number:
MFCD00002816 CAS Number: 156-54-7 EINECS: 205-857-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...