Glendale XC™ Laser Eyewear, Honeywell Safety
31-80102 31-80100 31-80103 31-80100 31-80108 31-80102 31-80111 31-80155 31-80155 31-80111 31-80103
89009-094EA 781.78 USD
89009-094 CA89009-090 89009-090 CA89009-104 89009-104 CA89009-100 89009-100 89009-098 CA89009-096 89009-096 CA89009-094
Glendale XC™ Laser Eyewear, Honeywell Safety
Eyewear features fully integrated lens dyes that provide superior protection and maximize visibility, while offering optimal attenuation by narrowing the absorption band to specific laser wavelengths
All models meet ANSI Z87+ impact and ANSI Z136.1 laser safety standards.
The dual 9-base wraparound polycarbonate lens provides extended peripheral vision and is available in a variety of tints. The lightweight frame is equipped with adjustable temple length and lens inclination features, as well as a soft elastomeric browguard and flexible fingers which cushion every point of contact with the face.
Ordering information: A detachable Rx insert frame is included with every pair of eyewear. Frame accepts inserts up to ±3 diopters. Storage case and hangcord are also included.