Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Uvex® GPT LOTG Series DVO® Laser Eyewear, Honeywell Safety

Supplier: HONEYWELL SAFETY PRODUCTS USA INC.
Uvex GPT LOTG Series DVO®
LOTG-532ALIGN LOTG-HENE
Providing protection against stray light from today's most common lasers, Uvex® LOTG eyewear is designed with a comfortable fit to encourage employees and lab workers to protect their eyes from incidental laser exposure

The Doubled Nd:YAG Align lens features an alignment filter for double-harmonic Nd:YAG lasers operating at 532nm (VLT of 65%).

The HeNe Align lens features an alignment filter for HeNe lasers operating at 633nm (VLT of 41%).

This over-the-glasses eyewear features a DVO® (diffuse viewing only) polycarbonate lens with Duoflex™ comfort cushions. Lenses are protected by antifog, anti-scratch, antistatic, anti-UV Uvextreme® AF coating. Can be worn over prescription eyewear or as a stand-alone spectacle. All DVO® laser eyewear is laser-inscribed across the lens with wavelength, corresponding OD, lot number, and part number for easy identification.



 

ORDER

Expand All / Collapse All
Filter Type Filter Wavelength Supplier No. VWR Catalog Number Unit Price Quantity
Doubled Nd:YAG Align 532 nm LOTG-532ALIGN 14217-744 Each Retrieving
HeNe Align 633 nm LOTG-HENE 14225-078 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty