MaxTag™ Immunoblotting Kits, Rockland Immunochemicals
KIA002 KIA001 KIA003
CAKIA003 CAKIA001 CAKIA002
MaxTag™ Immunoblotting Kits, Rockland Immunochemicals
These accurate and sensitive kits are ideal for western blotting or dot blotting methods
They offer flexibility with two unique substrates: Bright Blue TMB substrate and Stable Brown DAB substrate. Components are color-coded for error-free results. With MaxTag™ you can run up to 40 (10x10cm) immunoblots.
Ordering information: Complete with: Ultra Pure Tween® 20, IgG Bovine Serum Albumin, biotinylated anti-IgG antibody, enhanced Steptavadin-Peroxidase conjugate, TMB substrate and precipitating agent, DAB substrate tablets, hydrogen peroxide (H2O2) solution, reusable dropper bottles, control IgG, and instruction manual.