Puromycin dihydrochloride ≥98% (by TLC), Calbiochem®
540222-25MG 540222-100MG 540222-25MG 540222-100MG
80054-140EA 108.65 USD
CA80054-140 80054-140 CA80054-138 80054-138
Puromycin dihydrochloride ≥98% (by TLC), Calbiochem®
Puromycin dihydrochloride
Puromycin is a protein synthesis inhibitor.
  • Induces DNA fragmentation
  • Premature release of nascent polypeptide chains
An aminonucleoside antibiotic that acts as a prokaryotic and eukaryotic protein synthesis inhibitor. Resembles the aminoacyl-adenylyl terminus of aminoacyl-tRNA and competes for binding to the A-site of the large ribosomal subunit.
Inhibits translation by causing premature release of nascent polypeptide chains. Induces DNA fragmentation in thymocytes and in human HL-60 leukemia cells.