(-)-Epigallocatechin gallate ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
(-)-cis-3,4-Dihydro-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-1(2H)-benzopyran-3-yl Gallate
, (2R,3R)-5,7-Dihydroxy-2-(3,4,5-trihydroxyphenyl)chroman-3-yl 3,4,5-trihydroxybenzoate
, EGCG
M-Clarity™ quality level = MQ100
One of the main polyphenolic constituents of green tea that exhibits potent antitumor, anti-inflammatory, and antioxidant properties
Arrests cell cycling at G0/G1 phases and induces apoptosis in a dose-dependent manner. EGCG has been shown to inhibit PMA-induced skin thickening, and the activation of protein kinase C. Also inhibits the activation of ornithine decarboxylase, and activation of interleukin-1[alpha] mRNA and protein expression. Inhibits inducible nitric oxide synthase (iNOS) gene expression and enzyme activity and inhibits peroxynitrite-mediated formation of 8-oxodeoxyguanosine and 3-nitrotyrosine. Also strongly and directly inhibits telomerase.
Packaging: Packaged under inert gas
Formula:
C₂₂H₁₈O₁₁ MW: 458.38 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00075940 CAS Number: 989-51-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...