Acyclovir ≥98% (by HPLC), Calbiochem®, Millipore®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
2-amino-1,9-dihydro-9-((2-hydroxyethoxy)methyl)-6h-purin-6-one
, 1,9-dihydro-2-amino-9-((2-hydroxyethoxy)methyl)-6h-purin-6-on
, Acycloguanosine
, 2-Amino-9-((2-hydroxyethoxy)methyl)-1H-purin-6(9H)-one
M-Clarity™ quality level = MQ100
An antiviral agent
Inhibits the proliferation of NIH/3T3 fibroblastic cells transformed by either v-abl or bcr-abl. Synchronizes OM9; 22 leukemia cells at the G0/G1 phase of the cell cycle.
Formula:
C₈H₁₁N₅O₃ MW: 225.21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00057880 CAS Number: 59277-89-3 UN: 3316 ADR: 9, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...