Epoxomicin (synthetic) ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ100
324800-100UG
324800-100UG
80109-094EA
457.29
USD
CA80109-094
80109-094
Epoxomicin (synthetic) ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
Epoxomicin
An antitumor agent of microbial origin that acts as a potent, highly specific, and irreversible inhibitor of chymotrypsin-like (CT-L), trypsin-like (T-L), and peptidylglutamyl peptide hydrolyzing (PGPH) activities of the proteasome
Modifies the proteasomal catalytic subunits LMP-7, MECL1, and Z. Epoxomicin also displays anti-inflammatory activity in vivo.
Packaging: Packaged under inert gas
Formula:
C₂₈H₅₀N₄O₇ Storage Temperature: Freezer |
MDL Number:
MFCD03791061 CAS Number: 134381-21-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...