3-Aminobenzamide ≥97%, Calbiochem®
Supplier: MilliporeSigma
Synonyms:
m-Aminobenzamide
, INO-1001
165350-100MG
165350-100MG
80057-796EA
54.51
USD
80057-796
CA80057-796
3-Aminobenzamide ≥97%, Calbiochem®
3-Aminobenzamide
Antiproliferative agent that acts as an inhibitor of poly(ADP-ribose) polymerase (PARP). Has minimal effect on bacterial toxin-mediated ADP-ribosylation. Inhibitor of UV-induced apoptosis. Strongly inhibits K562 target cell killing by human effector natural killer cells.
Formula:
C₇H₈N₂O MW: 136.15 g/mol Boiling Pt: 341 °C (1013 hPa) Melting Pt: 111…115 °C |
MDL Number:
MFCD00007989 CAS Number: 3544-24-9 EINECS: 222-586-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...