Crews® Desperado™ Protective Eyewear, MCR Safety
Supplier: MCR Safety
Crews®
DS119 DS110AF DS112 DS110
80081-903EA 19.69 USD
80081-903 80081-892 80081-895 80081-900
Crews® Desperado™ Protective Eyewear, MCR Safety
Safety Glasses
These lightweight safety glasses feature black nylon temples and a frameless floating lens design that provides powerful protection in even the most rigorous circumstances.
Lenses are scratch-resistant and filter out 99% of UV radiation. Soft, secure gel nosepiece, bayonet temples, and nonslip gel temple sleeves provide a comfortable fit for extended wear. Glasses 80081-903 feature Duramass AF4® antifog lens coating.
Certifications: Meet or exceed ANSI Z87+ high impact safety standards.