Maxim™ Protective Eyewear, 3M™
Supplier: 3M
Maxim™
7010045584
47746-521EA 8.56 USD
47746-521
Maxim™ Protective Eyewear, 3M™
Safety Glasses
Maxim™ protective eyewear combines a broader range of view, adjustability, and comfort in safety glasses that are enjoyable to wear.
  • Adjustable features include 3-angle position lenses and temple lengths
  • Pantascopic lenses allow a greater range of view to broaden sightlines for the wearer
  • Comfort features include a dual-injected, cushioned brow bar, a soft nose bridge, and lightweight frames
  • DX™ anti-fog hard lens coating reduces the chances of fogging, scratching, or chemical damage
  • Sophisticated, contoured design offers an attractive appearance along with functional eye protection
  • Impact-resistant polycarbonate lenses absorb 99.9% UV rays and UVB up to 380 nm
The option to customize the safety glasses to the individual wearer makes them more pleasant to wear all day.
Eye protection is important to safeguard the user's eyes and maintain healthy vision. According to the Bureau of Labor Statistics, nearly three out of every five workers who suffered eye injuries were not wearing eye protection at the time of the accident. Wearing the correct safety eyewear for the task and for the unique needs of the worker is important. All 3M™ protective eyewear combines safety features with comfort enhancements.
These safety glasses are recommended for use in chipping, chiseling, drilling, grinding, machining, masonry, pouring/casting, power fastening, riveting, sanding, and sawing applications. Industries in which these safety glasses are commonly used include aviation, construction, food and beverage, general manufacturing, mining, oil and gas, primary metals, and transportation.
Certifications: ANSI Z87.1-2003. CSA Z94.3-2007.