JACKSON SAFETY® V50 CUDAS™ Safety Eyewear, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
JACKSON SAFETY®
14467
33001-228EA 17.61 USD
33001-228
JACKSON SAFETY® V50 CUDAS™ Safety Eyewear, KIMBERLY-CLARK PROFESSIONAL®
Safety Glasses
Stylish safety eyewear features a five-length adjustable temple for individual fit.
  • Frames made of durable, modified nylon
  • Optical-grade polycarbonate lens provides 99.9% UV protection
  • Choice of lens styles
For low-light indoor conditions, choose 33001-220 with amber lens. For antifog, anti-scratch and anti-static coating, choose 33001-218.
Six-base lens curve and folded-back lens design provide increased peripheral vision and protection.
Certifications: Meet ANSI Z87.1 standards. Meet high impact requirements of CSA Z94.3. Do not meet the sideshield requirements.