Would you like to log in again?
Nassau Rave™ Protective Eyewear, 3M™
Supplier: 3M OCCUPATIONAL HEALTH
Nassau Rave™ Nassau®
14451-00000-20 14460-00000-20 14459-00000-20 14450-00000-20
Nassau Rave™ protective eyewear features a wraparound lens that helps provide front and side protection
  • Wraparound lens helps provide front and side protection
  • Equipped with polymer frame and adjustable temples
  • Reliable choice in vision safety
Meets ANSI Z87.1 standard.
Equipped with a polymer frame and adjustable temples, it is a reliable choice in vision safety.
Certifications: ANSI Z87.1-2003. CSA Z94.3-2007.
ORDER
Expand All / Collapse All
Frame Material Style Frame Color Lens Coating Lens Tint Lens Color Supplier No. VWR Catalog Number Unit Price Quantity
HSE Plastics and Glass Half-Frame Black Anti-Scratch & Anti-Fog Clear Clear 14450-00000-20 55666-257 Case of 20 Retrieving
Each Retrieving
HSE Plastics and Glass Half-Frame Black Anti-Scratch & Anti-Fog IR 3.0 Shade 3.0 IR 14459-00000-20 89006-671 Case of 20 Retrieving
Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty